Ruthenium, black ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

12354-04 12354-09
CAAA12354-04EA 511.92 CAD
CAAA12354-04 CAAA12354-09
Ruthenium, black ≥99.9% (metals basis)
Ruthenium

Purity: 99.9% CAS No.: 7440-18-8 Odor: Odorless UN No.: 3089 MDL No.: MFCD00011207

Formula: Ru
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR