Sodium hexachloroosmate(IV) dihydrate Os ≥38,7%

Supplier: Thermo Scientific Chemicals
Warning

12176-01 12176-03 12176-06
CAAA12176-01EA 107.12 CAD
CAAA12176-01 CAAA12176-03 CAAA12176-06
Sodium hexachloroosmate(IV) dihydrate Os ≥38,7%
Disodium hexachloroosmate

CAS No.: 1307-81-9 Molecular Formula: Cl6Na2Os.2H2O Formula Weight: 484.93 (448.90anhy) Physical Form: Crystalline Sensitivity: Air and Moisture Sensitive Appearance: Red to brown Odor: Odorless MDL No.: MFCD00014234

Formula: Cl₆Na₂Os·2H₂O
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 389…393 °C
Density: 3,59 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014234
CAS Number: 1307-81-9
EINECS: 215-152-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR