Aluminium oxide activated basic ≥99.9995% Brockmann Grade I 58 angstroms

Supplier: Thermo Scientific Chemicals

11503-30 11503-A7
CAAA11503-30EA 111.23 CAD
CAAA11503-30 CAAA11503-A7
Aluminium oxide activated basic ≥99.9995% Brockmann Grade I 58 angstroms
Aluminium oxide activated basic

Uses: Suggested for the separation of basic amino acids, amines and other basic substances. Similar to neutral alumina, it will also adsorb hydrocarbons, aromatics and similar unsaturated compounds from organic solvents


CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: -60 Mesh Powder, S.A. 150m2/g Odor: Odorless Melting Point: 2045° MDL No.: MFCD00003424


Suggested for the separation of basic amino acids, amines and other basic substances. Similar to neutral alumina, it will also adsorb hydrocarbons, aromatics and similar unsaturated compounds from organic solvents

Formula: Al₂O₃
Melting Pt: 2045 °C
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
UN: 1325
ADR: ,
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR