Aluminium oxide activated acidic Brockmann Grade I 58 angstroms

Supplier: Thermo Scientific Chemicals

11501-30 11501-A1 11501-A7
CAAA11501-30EA 56.09 CAD
CAAA11501-30 CAAA11501-A1 CAAA11501-A7
Aluminium oxide activated acidic Brockmann Grade I 58 angstroms
Aluminium oxide activated acidic

Uses: Suggested for the separation of both inorganic anions and acidic organic molecules such as acidic amino acids, aromatic acids and carboxylic acids


CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: -60 Mesh Powder, S.A. 150m2/g Odor: Odorless Melting Point: 2045° MDL No.: MFCD00003424


Suggested for the separation of both inorganic anions and acidic organic molecules such as acidic amino acids, aromatic acids and carboxylic acids

Formula: Al₂O₃
MW: 101.96 g/mol
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR