Glass Stirring Shafts, 19 mm, Ace Glass
Supplier: Ace Glass
8076-41 8076-10 8076-40 8076-42 8076-43 8076-44 8076-45 8076-46 8076-48 8076-05 8076-07 8076-39 8076-03
71001-570EA 925.81 CAD
71001-570 89061-990 89061-992 89061-994 89061-996 89061-998 89062-000 89062-002 89062-004 89062-006 89062-010 89235-692 10099-470
Glass Stirring Shafts, 19 mm, Ace Glass
Stirrers Stirring Shafts
Polished glass stirring shafts, 19mm O.D
Available in solid and hollow styles, and in various lengths. Join PTFE stir blades to button type shafts, or join PTFE multi-blade, PTFE single blade, PTFE turbine, and PTFE multi-paddle agitators to either knob or drilled style shafts. Recommended for use with ACE mechanical seal and PTFE debris-free bearings. Use ACE Flex-Grip chucks to join the shaft to a stirring motor.