Joints, Medium Length, Ace Glass Incorporated
Supplier: Ace Glass
7585-16 7585-28 7585-46 7585-02 7585-04 7585-08 7585-10 7585-14 7585-20 7585-22 7585-26 7585-32 7585-34 7585-38 7585-40 7585-44 7585-50 7585-52 7585-56 7585-58 7585-65 7585-67
89059-540EA 41.8 CAD
89059-540 89059-542 89059-548 89059-552 89059-554 89059-556 89059-558 89059-560 89059-564 89059-566 89059-568 89059-572 89059-574 89059-576 89059-578 89059-580 89059-584 89059-586 89059-588 89059-590 89059-592 89059-594
Joints, Medium Length, Ace Glass Incorporated
NS Joint Sleeves and Connectors Joint Sleeves
The outer diameter of both joint and tube are approximately the same size, allowing for ease in clamping
All joints are manufactured from borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable