Thermowell with [ST] Joint, Ace Glass
Supplier: Ace Glass
6500-01 6500-02 6500-03 6500-04 6500-05 6500-06 6500-08 6500-10 6500-12 6500-20
89055-018EA 113.79 CAD
89055-018 89055-020 89055-022 89055-024 89055-026 89055-028 89055-030 89055-032 89055-034 89055-036
Thermowell with [ST] Joint, Ace Glass
Laboratory Reactor Systems
Thermowell features a 14/35 [ST] joint for use with spherical or cylindrical reaction flasks.
The length measurement (A as illustrated) listed below indicates the distance below the joint to the bottom of the tube. All Thermowells are fabricated from borosilicate glass.