Tweezers, Straight Taper Relieved Tips, Style 4, Excelta Corp®
Supplier: Excelta
4-SA 4-SA-PI 4-SA-SE
63042-368EA 140.1 CAD
63042-368 63042-372 63042-374 89411-846
Tweezers, Straight Taper Relieved Tips, Style 4, Excelta Corp®
Forceps
Designed for microscopy, the tapered tips offer excellent visibility under a scope.
  • Specifically made for microscopy
  • Available in three different materials
  • Three different quality grades available
  • Unsurpassed visibility under a scope
  • Three quality grades available
The 'Style 4' straight tapered ultra-fine pointed tweezers is primarily used for microscopy. The overall length is 4.25". These are also availabe with ergonomic handles (ET). We offer 3 different qualities. The higher the quality, the more precise the tips. They are made of stainless anti-magnetic steel.