Powder Funnels, Offset, Ace Glass Incorporated
Supplier: Ace Glass
7237-09 7237-11 7237-15 7237-19 9489-20
89058-628EA 229.26 CAD
89058-628 89058-630 89058-632 89058-634 89074-942
Powder Funnels, Offset, Ace Glass Incorporated
Funnels Powder Funnels
Used for pouring liquids or powders into multi-neck flasks
Has a standard taper inner joint at bottom. Fabricated from borosilicate glass.