Protector® Premier® Laboratory Hoods, Labconco®
Supplier: Labconco
100400000 100400002 100400020 100400021 100400040 100400042 100400060 100400061 100400070 100500000 100500002 100500020 100500021 100500040 100500042 100500060 100500061 100500070 100600000 100600002 100600020 100600021 100600040 100600042 100600060 100600061 100600070 100800000 100800002 100800020 100800021
89232-730EA 17008.7 CAD
89232-730 89232-732 89232-734 89232-736 89232-738 89232-740 89232-742 89232-744 89232-746 89232-748 89232-750 89232-752 89232-754 89232-756 89232-758 89232-760 89232-762 89232-764 89232-766 89232-768 89232-770 89232-772 89232-774 89232-776 89232-778 89232-780 89232-782 89232-784 89232-786 89232-788 89232-790
Protector® Premier® Laboratory Hoods, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Labconco Protector® Premier® fiberglass fume hoods have a one-piece liner of specially-formulated, fiberglass-reinforced polyester, which offers corrosion and fire resistance and easy clean up.
These by-pass hoods feature the largest unobstructed sightline available (37.5" from work surface to header panel). Other features include Eco-Foil™ air foil that corrects natural turbulence while keeping cords out of the way, dry powder-coated steel exterior, safety glass sash, and molded fiberglass exhaust collar. Standard hoods include pre-wired LED lighting rated for 50000 hours. Explosion-proof (EP) models include incandescent lighting (bulb not included).
Hoods are available with a built-in standard fiberglass blower, or an explosion-proof fiberglass blower, or without a blower (remote blower sold separately).
There are no seams where deterioration can occur or where contaminants can collect. These general purpose hoods are ideal for corrosive fumes but should not be used with perchloric acid, or radioactive or biological hazards.
Certifications: All models are ETL listed. 230V models are CE listed.
Ordering information: Customize your hood with optional service fixtures that accommodate air, gas, vacuum or water lines. Complete your fume hood with a variety of accessories including blowers, work surfaces, storage cabinets, fire extinguishers, and air flow monitors (sold separately).