Stainless Steel Work Tables, Wall Mounted, Eagle MHC
Supplier: Eagle Group
WT304814304SEM WT306014304SEM WT307214304SEM
89207-564EA 5323.23 CAD
89207-564 89207-566 89207-568
Stainless Steel Work Tables, Wall Mounted, Eagle MHC
Furniture Tables and Workbenches
14 gauge Type 304 stainless steep construction.
  • Box marine counter edge
  • Weight capacity, 200 lbs.
14-gauge brackets with hole for electrical wire run wall mounted for environments witih frequent wash downs.