VWR® Modular Workstations, W×D×H 60×30×36"
Supplier: VWR International
VF-B603036E VF-B603036EB VF-B603036F VF-B603036L VF-B603036P VF-B603036PB
VF-B603036EEA 4988.39 CAD
VF-B603036E VF-B603036EB VF-B603036F VF-B603036L VF-B603036P VF-B603036PB
VWR® Modular Workstations, W×D×H 60×30×36"
Furniture Tables and Workbenches
VWR® modular work benches are designed for a wide range of applications in laboratory and industrial environments including biotech, pharmaceutical, analytical, material handling, distribution, and assembly.
  • Can be custom configured
  • Shipped fully assembled
  • Includes a variety of attachments
Standard work benches handle 1000lbs
The VWR® Basic Bench includes 4 standard leveler bench feet, gussets and light neutral powder coat of 16 gauge 1.5x1.5” steel frame. Shipped fully assembled. Also available in 48" and 72" widths.
of evenly distributedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable