VWR® Modular Workstations, W×D×H 48×30×36"
Supplier: VWR International
VF-B483036E VF-B483036EB VF-B483036F VF-B483036L VF-B483036P
VF-B483036EEA 4394.26 CAD
VF-B483036E VF-B483036EB VF-B483036F VF-B483036L VF-B483036P
VWR® Modular Workstations, W×D×H 48×30×36"
Furniture Tables and Workbenches
VWR® modular work benches are designed for a wide range of applications in laboratory and industrial environments including biotech, pharmaceutical, analytical, material handling, distribution, and assembly.
  • Can be custom configured
  • Shipped fully assembled
  • Includes a variety of attachments
Standard work benches handle 1000lbs
The VWR® Basic Bench includes 4 standard leveler bench feet, gussets and light neutral powder coat of 16 gauge 1.5x1.5” steel frame. Shipped fully assembled. Also available in 60" and 72" widths.
of evenly distributed weight but can be configured to handle loads of 5000lbs. or more. Tables can be custom configured with numerous accessories including, 8" adjustable leg kits (for ADA compliance), locking casters, heavy-duty 2x2 steel frame, upper and lower shelves with pencil lips, backsplashes, lighting, bin rails, power strips, ESD tops, and steel drawers.
Ordering information: Select work bench size followed by type of work surface (epoxy, plastic laminate, ESD, phenolic). Next add appropriate accessories to the bench to complete your configuration. If you need something not on the price list, then please contact your Avantor sales representative. Exact frame dimensions depend upon reveal and thickness of work surface to be added.