VWR® Modular Workstations, W×D×H 60×30×30"
Supplier: VWR International
VF-B603030E VF-B603030EB VF-B603030F VF-B603030L VF-B603030P VF-B603030PB
VF-B603030EEA 4893.24 CAD
VF-B603030E VF-B603030EB VF-B603030F VF-B603030L VF-B603030P VF-B603030PB
VWR® Modular Workstations, W×D×H 60×30×30"
Furniture Tables and Workbenches
VWR® modular work benches are designed for a wide range of applications in laboratory and industrial environments including biotech, pharmaceutical, analytical, material handling, distribution, and assembly.
  • Can be custom configured
  • Shipped fully assembled
  • Includes a variety of attachments
Standard work benches handle 1000lbs
The VWR® Basic Bench includes 4 standard leveler bench feet, gussets and light neutral powder coat of 16 gauge 1.5x1.5” steel frame. Shipped fully assembled. Also available in 48" and 72" widths.
of evenly distributedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable