Receiver, Moisture Test, Barrett-Type, Ace Glass
Supplier: Ace Glass
7745-02 7745-04 7745-21 7745-26 7745-102 7745-202 7745-104 7745-206
89060-342EA 580.48 CAD
89060-342 89060-344 89060-346 89060-348 89082-862 89082-864 89082-866 89082-868
Receiver, Moisture Test, Barrett-Type, Ace Glass
Distillation Components Distillation Receivers
This moisture test receiver is used to determine the water content in petroleum and bituminous material
It is supplied with [ST] 24/40 joints, and with a stopcock to empty the measuring tube during the determination. The 10mL size is subdivided in 0.1mL from 0–1mL, 0.2mL from 1–10mL; 20mL size in 0.1mL from 0–1mL, 0.2mL from 1–20mL. Fabricated from borosilicate glass.
Ordering information: Available with 2mm bore glass, or with 1:5 solid PTFE stopcock plug. Available with jacketed or unjacketed side arm. Jacketed side arm version helps prevent condensation and facilitates improved performance.