Scissors, Miniature Self-Opening, Precision, Excelta Corp®
Supplier: Excelta
346 346A 346B 349 349A 349B 366 367
63041-984EA 99.79 CAD
63041-984 63041-986 63041-998 63042-000 63042-002 63042-004 63042-032 63042-034 63042-036 76471-556
Scissors, Miniature Self-Opening, Precision, Excelta Corp®
Scissors
Self-opening miniature scissors are ideal for delicate cutting applications where space is at a premium.
  • Miniature for cutting under a scope
  • Cleanroom safe, stainless steel
  • Self-opening/spring loaded for ease of use
These two star self-opening precision scissors with precision cutting blades are ideal for delicate cutting applications. The stainless steel blade is hardened to Rockwell C-50. They are open at rest and spring-loaded.