Thermometers, PTFE Coated for Safety, Ace Glass
Supplier: Ace Glass
8283-10 8283-15 8283-11 8283-03
89084-760EA 141.71 CAD
89084-760 89084-762 89084-766 89084-780
Thermometers, PTFE Coated for Safety, Ace Glass
Thermometers Analog Thermometers
These double-safe thermometers have a PTFE coating
Black liquid filled, yellow back, lead free glass. In the event of breakage, the material is contained by the PTFE until it can be safely disposed of. These thermometers are individually serialized and N.I.S.T. traceable.
Order Now
SPECIFICATIONS
Environmentally Preferable