Brushes, Scratch, Replaceable Tips, Excelta Corp®
Supplier: Excelta
264 265 267 268 266
63041-704EA 66.9 CAD
63041-704 63041-710 63041-714 63041-730 63041-738 63041-742 75880-970
Brushes, Scratch, Replaceable Tips, Excelta Corp®
Brushes
Used for cleaning and polishing, these dial-out bristles can be stiff or soft, depending on how far they protrude from the handle.
  • Replaceable bristles
  • Bristles 'dial-out' for different stiffness
  • Four different bristle materials
These scratch brushes feature replaceable bristles with a plastic handle. We offer bristles of brass, steel, fiberglass and nylon. Simple twist the collar and the bristles appears. It can be dialed out, to your preference, based on how soft or hard you want the bristles.
Delivery information: The kit includes one handle andMore Product Information