Tweezers, Straight Fine Tips, Style 1, Excelta Corp®
Supplier: Excelta
1-PI 1-S 1-SA 1-SA-PI 1-SA-SE 1-S-SE 1-SA-ET
63041-404EA 66.49 CAD
63041-404 63041-406 63041-408 63041-412 63041-414 63041-416 89411-818
Tweezers, Straight Fine Tips, Style 1, Excelta Corp®
Forceps
The style 1 tweezers are designed to work with medium magnification (scope).
  • Design is excellent for work under scope
  • Available in 3 different quality grades
  • Available in 3 different materials depending on application
The 'Style 1' Series tweezers is ideal for delicate applications. They feature fine straight points and are designed for bending and handling very fine wires and components. We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) orMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable