Tweezers, Flat Duck-Bill Tip, Style 2A, Excelta Corp®
Supplier: Excelta
2A 2AB-N 2AB-SA-PI 2AB-SA-SE 2A-S 2A-SA 2A-SA-PI 2A-SA-SE 2A-S-SE 2A-SA-PI-ET F-SA F-SA-PI 2A-SA-PI-TC15 2A-SA-TC15 2A-SA-TC30 2A-SA-R
63041-862EA 150.24 CAD
63041-862 63041-864 63041-866 63041-868 63041-872 63041-874 63041-876 63041-880 63041-882 63041-884 89107-634 94024-406 94024-408 75880-524 75880-526 75880-528 76471-458 76471-460
Tweezers, Flat Duck-Bill Tip, Style 2A, Excelta Corp®
Forceps
The 2A series is ideal for handling SMD components and labels.
  • Excellent for handling labels/smd components
  • Available in 3 different materials
  • Three different quality grades
  • Available with ergonomic handles
  • Teflon coated tips available, soft handling
The 'Style 2A' Series tapered flat round tipped tweezer is designed for handling labels or SMD components. The different options offered are ergonomic handles or teflon coated tips (to prevent scratching).depending on your application. We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic).