Pressure Bearing with Ace-Threds, 10 mm, Ace Glass Incorporated
Supplier: Ace Glass
8044-07 8044-13 8044-24 8044-55
89061-790EA 138.22 CAD
89061-790 89061-792 89061-794 89061-796
Pressure Bearing with Ace-Threds, 10 mm, Ace Glass Incorporated
Stirrers Stirring Shafts
Glass bearings feature a #15 Ace-Thred at each end and glass tubing between
Designed for use with 10mm O.D. stirring shafts. A PTFE coupling, with internal O-Ring seal, connects the bottom of the bearing to either a #15 or a #25 Ace-Thred on vessels such as a pressure reactor head or one-piece pressure reactors. The top bushing also has an internal O-Ring, for an additional seal on the shaft to allow pressure reactions. Maximum operating speed is 600rpm. Coupling and bushing are each supplied with O-Rings. Glass components are fabricated from borosilicate glass.
Ordering information: Items can be purchased complete with bearing, coupling (89052-638 for #15–#15 or 89052-684 for #15–#25), and bushing, or individual components may be purchased separately. For replacement O-Rings for #15–#15 coupling order 89061-012 (external, size -210) and 89060-984 (internal, size -110); for #15–#25 coupling, order 89061-012 and 89061-028 (external, size -210 and -220) and 89060-984 (internal, size -110); for #15 bushing, order 89061-012 (external, size -210) and 89060-984 (internal, size -110).