Cutters, Standoff Shear, Small Frame, Excelta
Supplier: Excelta
500-106-US 509-42A-US 530CE-US 530E-15-US 530E-1-US 530E-4-US 530E-5-USL 530EA-US 530E-US 549E-US 530E-USL
63042-402EA 977.12 CAD
63042-402 63042-498 63042-526 63042-530 63042-532 63042-538 63042-540 63042-542 63042-546 63042-580 75880-926
Cutters, Standoff Shear, Small Frame, Excelta
Cutters
Designed for work with printed circuit boards, these tools cut leads that protrude from the board and reduce shock on delicate solder connections or components.
  • Anti-shock trimming of leads to specific length
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
This popular anti-shock shear cutter ensures that each and every lead is cut precisely to the desire length. There are many different configurations of this tool. Those with 'CUSTOM' suffixes require the buyer to specificy the desired lead length. They feature an ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. These tools are made of carbon tool steel and have flame hardened cutting edges.