Tweezers, Surface Mount, Excelta Corp®
Supplier: Excelta
111-SA 121-SA 122-SA 123-SA 124-SA 125-SA 126-SA 128A-SA 102A-SA 103-SA 104-SA 109-SA 110-SA
63041-132EA 251.16 CAD
63041-132 63041-192 63041-194 63041-196 63041-198 63041-200 63041-202 89411-880 75880-538 75880-540 75880-542 76471-526 76471-528
Tweezers, Surface Mount, Excelta Corp®
Forceps
These tools are specifically designed to handle various electronic Surface Mount Devices.
  • Specifically designed for specific applications
  • Made of anti-magnetic stainless steel
  • Fluted handles for better control
These S.M.D. paddle tip tweezers will firmly hold many types of SMD components. They are made of anti-magnetic Neverust® stainless steel and features serrated handles for better contol.
Order Now
SPECIFICATIONS
Environmentally Preferable