Tweezers, Carbofib Soft Replaceable Tip, Excelta Corp®
Supplier: Excelta
159D-RT 159-RT 159A-RT 159E-RT 159C-RT 159-RT-ET 159F-RT 159G-RT
16003-848EA 120.46 CAD
16003-848 63041-264 63043-714 63046-724 94024-118 89411-864 75880-634 75880-636 75880-844 76471-534 76471-536 76471-540
Tweezers, Carbofib Soft Replaceable Tip, Excelta Corp®
Forceps
Carbon Fiber tipped tweezers prevent nicking and marring of delicate items while maintaining high rigidity and tensil strength.
  • Replaceable tips
  • Available in a multitude of tip sizes
  • Temperature resistant to 249 °F
  • Conductive
  • Will not scratch or mar parts
  • Excellent tensile and flexural strength
These soft replaceable tip tweezer boasts Carbofib™ tips. The Carbofib™ tips are a polymer based material with 30% carbon fibers added (conductive 10² ohms/sq). Carbofib™ has very high rigidity, fatigue and creep resistance. The bodies are made of anti-magneticMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable