Tweezers, Straight Fine Taper Tips, Style 2, Excelta Corp®
Supplier: Excelta
2-S 2-SA 2-SA-SE 2-S-SE 2-SA-ET
63041-890EA 134.11 CAD
63041-890 63041-892 63041-898 63041-900 89411-826
Tweezers, Straight Fine Taper Tips, Style 2, Excelta Corp®
Forceps
These long taper tips offer better visibility and works well under low magnification.
  • Tapered tips allow for greater visibility/accessibility
  • Three different qualities
  • Three different materials to choose from
The 'Style 2' Series is a straight tapered fine precision pointed tweezers. This series offers better visibility and works well under low magnification. We offer 2 different qualities. The higher the quality, the more precise the tips. They are offered in stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic). It isMore Product Information