SP Bel-Art Check Valves, PP, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H19717-0000 H19715-0000 H19716-0000
16126-432PK 160.8 CAD
16126-432 16126-634 16126-636
SP Bel-Art Check Valves, PP, Bel-Art Products, a part of SP
Valves Vacuum Valves
Polypropylene plastic valve with neoprene valve disk stops the reverse flow of liquid when pressure is interrupted.
  • Polypropylene construction
  • Autoclavable
  • Neoprene valve disk
Useful when connecting a vacuum line to a filter flask. Recommended maximum pressure 20psi (1034 Torr). Autoclavable.
Order Now
SPECIFICATIONS
Environmentally Preferable