Polypropylene Buckets, Contec®
Supplier: CONTEC, INC. MX
UBKT0003 UBKT0005 UBKT0006 UBKT0001 UBKT0002
89198-842CS 128.87 CAD
89198-842 89198-846 89198-848 94025-806 94025-808
Polypropylene Buckets, Contec®
Buckets
Contec® bucket systems are available in a variety of size options for small and large critical environments. Replacement buckets are available for the polypropylene and stainless systems.
  • Replacement buckets for polypropylene and stainless bucket systems
  • For use with 3.9, 5, 6.5, and 7 gal. bucket systems
  • Autoclavable
  • Available in a variety of colors
These buckets are autoclavable and work with a variety of disinfectants and solvents. Wringer options are available based on bucket size.