E.Z.N.A.® Tissue DNA Extraction Systems, Omega Bio-tek
Supplier: Omega Bio-Tek
E.Z.N.A®
D3096-02 D1196-00 D3396-02 D3396-00 D3096-00 D3396-01 D3096-01 D3396-00 D1196-01 D3096-00 D3396-02 D3096-02 D1196-00 D3396-01 D1196-01 D3096-01
CA101318-988EA 476.39 CAD
CA101318-988 101319-016 CA101414-370 CA101318-986 101319-018 CA101414-382 CA101318-948 101318-990 CA101319-016 101318-986 CA101319-018 101414-370 101414-382 101318-988 101318-948 CA101318-990
E.Z.N.A.® Tissue DNA Extraction Systems, Omega Bio-tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Omega Bio-tek® provides multiple options for genomic, viral, and mitochondrial DNA extraction from cultured cells, buccal swabs, mouse tail snips, whole blood, buffy coat, and a variety of animal tissue
The SQ Tissue DNA Kit is a unique scalable system that permits the isolation of high molecular weight DNA from variable starting sample amounts. The tissue is lysed with a detergent-based buffer and proteins are precipitated out with salt solution. The released DNA is purified and concentrated via alcohol precipitation.
The Mag-Bind® Tissue DNA systems are used for preparation of genomic, viral, and mitochondrial DNA from animal tissue, buccal swabs, mouse tails, whole blood, buffy coat, and cultured cells using paramagnetic beads. Magnetic beads can be used in 24, 96, or 384-well formats to fit your sample size, with the flexibility to process manually or with automation. By eliminating retention volume commonly seen with filter-based applications, magnetic beads allow for higher recovery and optimal yield. Magnetic stands can be added to liquid handlers which utilize the SBS format and can be programmed for your application allowing simultaneous processing of multiple plates. The technology allows for higher purity and flexible elution volumes. The Mag-Bind® Tissue DNA Kits are compatible with open formatted instruments such as Hamilton Nimbus, Starlet, Star Plus, Tecan Freedom Evo®, Beckman Coulter Biomek® 2000, 3000, NX, FX, and Thermo Kingfisher® Flex and Kingfisher® Duo.
The E.Z.N.A.® Tissue DNA Kit uses silica membrane technology accompanied by a specialized buffer system to purify DNA from 30mg of tissue or 5x10⁶ cells. The E-Z 96 silica plates can be used with a vacuum manifold, centrifugation, or automated vacuum systems. It is scalable depending on the size of the sample. Up to 10mg of tissue or 2.5 x 10⁶ cells can be purified using the MicroElute® Tissue DNA Kit elution volume.