SP Bel-Art Drum and Carboy Funnels, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H14712-0350 H14712-0200 H14712-0250
47750-840EA 149.54 CAD
47750-840 47750-836 47750-838
SP Bel-Art Drum and Carboy Funnels, Bel-Art Products, a part of SP
Funnels Filtration Funnels Drum Funnels
Large funnels are ideal for filling drums, barrels, tanks and carboys providing ample volume and fast flow.
  • Natural polypropylene construction
  • Autoclavable
  • Large funnel provides ample volume and fast flow
  • Handle in some models adds stability
The two smaller sizes have a handle at the rim for added stability. Molded of natural polypropylene, funnels are steam autoclavable at 121ºC (250ºF).