VK Sponge, Contec®
Supplier: Contec
VKST0003 VKSPONGE-1
33502-266CS 1497.29 CAD
33502-266 68310-407
VK Sponge, Contec®
Wipes Sponges
The VertiKlean® Sponge is a disposable cleanroom sponge that features high sorbency polyester foam laminated to a Polynit wiping material with excellent abrasion resistance.
  • High sorbency polyester foam makes sponges very sorbent
  • Textured polyester surface for more aggressive cleaning tasks
  • The sponge is laser cut to completely seal the edges
Designed for general cleaning and wiping, they are ideal for use in the laboratory. Autoclavable or supplied as a sterile, ready-to-use product.
Packaging: Non sterile sponges are packaged 8/bag, 3 bags/innerMore Product Information