Keyed Quick Connect Plug, WaterSaver Faucet
Supplier: WaterSaver Faucet
QCD-B-3M-K1 QCD-B-3M-K2 QCD-B-3M-K3 QCD-B-3M-K4 QCD-B-3M-K5 QCD-B-3M-K6 QCD-B-3M-K7 QCD-B-3M-K8
89176-966EA 141.22 CAD
89176-966 89176-968 89176-970 89176-972 89176-974 89176-976 89176-978 89176-980
Keyed Quick Connect Plug, WaterSaver Faucet
Fittings and Connectors
Plugs have an internal shut-off valve that closes when the valve is disengaged.
Internal valve components are stainless steel to assure compatibility with service to assure durable performance.
Certifications: CSA certified for natural gas.