Keyed Quick Connect Body, WaterSaver Faucet
Supplier: WaterSaver Faucet
QCFL-B-3M-K1 QCFL-B-3M-K2 QCFL-B-3M-K3 QCFL-B-3M-K4 QCFL-B-3M-K5 QCFL-B-3M-K6 QCFL-B-3M-K7 QCFL-B-3M-K8
89176-950EA 248.14 CAD
89176-950 89176-952 89176-954 89176-956 89176-958 89176-960 89176-962 89176-964
Keyed Quick Connect Body, WaterSaver Faucet
Fittings and Connectors
The quick connect keying system is based upon the inner and outer diameters of the sleeves on the body and plug
Quick connect bodies and plugs color coded to designate the service and to facilitate matching the correct body and plug.
Certifications: CSA certified for natural gas.