Anti-RAGE Rabbit Polyclonal Antibody

Supplier: Genetex
GTX23611
89367-558EA 938.19 CAD
89367-558
Anti-RAGE Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to RAGE

Recommended Starting Dilutions: For ICC/IF: Use at a dilution of 1:200. For WB: 1:500-1:10000. For IHC-Fr: Use at a concentration of 1-2 ug/ml. For IHC-P: 1:100-1:1000. Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: RAGE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR