Anti-MRP4 Rat Monoclonal Antibody [clone: M4I-80]

Supplier: Genetex
GTX15598
89367-422EA 1188.33 CAD
89367-422
Anti-MRP4 Rat Monoclonal Antibody [clone: M4I-80]
Antibodies
May be an organic anion pump relevant to cellular detoxification.

ICC: Use at a dilution of 1/20 - 1/50. IHC-Fr: Use at a dilution of 1/20. WB: Use at a dilution of 1/20 - 1/50. Predicted molecular weight: 150 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MRP4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR