Anti-TUBB Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX15568
89351-196EA 983.96 CAD
89351-196
Anti-TUBB Rabbit Polyclonal Antibody
Antibodies
Tubulin is the major building block of microtubules. This intracellular cylindrical filamentous structure is present in almost all eukaryotic cells. Microtubules function as structural and mobile elements in mitosis, intracellular transport, flagellar movement, and in the cytoskeleton.

IHC-P: Use at a dilution of 1/200 for 10 minutes at RT. IP: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 55 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TUBB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR