Anti-GFP Chicken Polyclonal Antibody

Supplier: Genetex
GTX13970
89359-920EA 1270.98 CAD
89359-920
Anti-GFP Chicken Polyclonal Antibody
Antibodies
The jellyfish Aequorea victoria contains green fluorescent protein (GFP) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organisms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcellular compartments where these proteins are targeted. GFP technology has revealed considerable new insights into the physiological activities of living cells.

Type: Primary
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Other
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR