Septa, Flat, Microscale, Ace Glass Incorporated
Supplier: Ace Glass
8787-41 8787-42 8787-43 8787-55 8787-58 8787-40
89064-074CS 133.33 CAD
89064-074 89064-076 89064-078 89064-080 89064-082 89064-072
Septa, Flat, Microscale, Ace Glass Incorporated
Septa
Replacement flat septa are designed for use with microscale reaction vials, or any other V-Bottom reaction vials.
These septa are constructed of white silicone rubber, bonded with a white PTFE facing. They will resist coring when punctured by a syringe needle. All septa are 3 mm thick.