Bubbler, Mineral Oil, Adjustable, Ace Glass Incorporated
Supplier: Ace Glass
8762-03 8762-07 8762-14
89063-990EA 154.21 CAD
89063-990 89063-994 89063-998
Bubbler, Mineral Oil, Adjustable, Ace Glass Incorporated
Bubblers
This mineral oil bubbler has a completely adjustable 7mm inner tube
The tube has a pulled tip, and it is secured to the vessel via #7 Ace-Thred. Volume of the bubbler is approximately 40mL. Tapered bottom of vessel allows for use with small volumes. A #7 Ace-Thred nylon bushing (89051-122) is supplied, along with a FETFE O-ring. Manufactured from borosilicate glass.
Ordering information: Available as a complete unit with vessel, inner tube, and bushing/O-ring, or the components may be purchased separately. For bushing with O-ring (only), see item 89051-122. An optional inner tube withMore Product Information