AdjusTable® Workstation, Stainless Steel Perforated Top, Stainless Steel Base, Eagle MHC™
Supplier: Eagle Group
AP2436T AP2448T AP2460T AP3036T AP3048T AP3060T
58202-852EA 4841.7 CAD
58202-852 58202-854 58202-856 58202-858 58202-860 58202-862
AdjusTable® Workstation, Stainless Steel Perforated Top, Stainless Steel Base, Eagle MHC™
Furniture Tables and Workbenches
Type 304 perforated brushed stainless steel top.
  • 14 gauge worksurface with ¾" perforations on 1" centers
  • Stainless steel base feature four 33" stainless posts
  • One stainless wire shelf and stainless C-frame
Load capacity: 600 lbs.