E.Z.N.A.® Endo-Free Plasmid Maxi Kit, Omega Bio-tek®

Supplier: OMEGA BIO-TEK CA

D6926-03 D6926-04 D6926-00
CA101319-342EA 947.56 CAD
CA101319-342 CA101319-360 CA101319-388 CA101414-406
E.Z.N.A.® Endo-Free Plasmid Maxi Kit, Omega Bio-tek®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

The Endo-Free Plasmid Maxi Kits provide an efficient method for the large-scale isolation of endotoxin-free plasmid DNA


  • Endotoxin-Free
  • Format Maxi


Following lysis, endotoxins are removed from the cleared lysate with the addition of ETR Solution. Plasmid DNA is then further purified by adsorbing to the HiBind® silica membrane. An easy wash step is followed by the elution of the bound plasmid DNA in elution buffer. Up to 200µg of high-quality plasmid DNA can be purified from 30–50mL of overnight culture (D6915), and up to 1mg from 100–200mL of overnight culture (D6926). Purified DNA samples are ready to be used in downstream applications such as automated fluorescent DNA sequencing, restriction enzyme digestion, transfection of mammalian cells, cloning, and PCR amplification.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR