E-Z 96® Total RNA Kit I, Omega Bio-tek®

Supplier: OMEGA BIO-TEK CA

R1034-02 R1034-00
CA101319-182EA 2304.7 CAD
CA101319-182 CA101319-354 CA101414-820
E-Z 96® Total RNA Kit I, Omega Bio-tek®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

Based on the same principle of the Total RNA Kit I, the E-Z 96® Total RNA Kit is designed to isolate total RNA from cells and soft tissues in a 96-well plate format


Up to 5 x 10^5 cells and 10mg of soft tissues can be processed in each well of the E-Z 96® RNA plate. This kit allows single or multiple simultaneous processing of samples in less than 60 minutes. There is no need for phenol/chloroform extractions, time-consuming steps such as CsCl gradient ultracentrifugation, or precipitation with isopropanol. Purified RNA is suitable for most downstream applications such as RT-PCR, qPCR, differential display, and microarrays.

Order Now

SPECIFICATIONS

Description E-Z 96® Total RNA Kit
Environmentally Preferable
Application
Target molecule RNA
Refine target molecule Total RNA
Sample type Cultured cells,Human/animal cells and tissues
Method / Format Multiwell plate
Prep size Mini

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR