Tetrahydrofuran-[D₈] ≥99.5% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Deuterated Tetrahydrofuran, Octadeuterotetrahydrofuran, Tetrahydrofuran-[²H₈], THF-d8, THF-D8, Tetrahydrofuran-d8

M-Clarity™ quality level = MQ200

1.13364.0001 1.13364.0009 1.13364.0010
CA1.13364.0001EA 120 CAD
CA1.13364.0001 CA1.13364.0009 CA1.13364.0010
Tetrahydrofuran-[D₈] ≥99.5% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Tetrahydrofuran-[D8]

MagniSolv™ deuterated solvents provide:
• Reliable results in the NMR-spectra by excellent chemical purity and highest isotopic enrichment.
• Reliable deuteration degrees.
• Low residual water content.
• Determination of water content in two ways.

Formula: C₄D₈O
MW: 80.04 g/mol
Boiling Pt: 65 °C (1013 hPa)
Melting Pt: –108 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: –17 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00044238
CAS Number: 1693-74-9
EINECS: 216-898-4
UN: 2056
ADR: 3,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR