N,N-Dimethylformamide-[D₇] ≥99.5% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Heptadeutero-N,N-dimethylformamide, N,N-Di[2H3]methyl[2H]formamide, N,N-Dimethylformamide-d7, DMF-D7, DMF-d7, N,N-Dimethylformamide-[²H₇], Formdimethylamide-D7, Formic acid dimethylamide-D7
M-Clarity™ quality level = MQ200
1.11656.0001
1.11656.0009
CA1.11656.0001EA
234
CAD
CA1.11656.0001
CA1.11656.0009
N,N-Dimethylformamide-[D₇] ≥99.5% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
N,N-Dimethylformamide-[D7]
MagniSolv™ deuterated solvents provide reliable results in the NMR-spectra with extremely low residual water, excellent chemical purity and the highest isotopic enrichment.
Formula:
DCOON(CD₃)₂ MW: 80.04 g/mol Boiling Pt: 152 °C (1013 hPa) Melting Pt: –60 °C Density: 1.03 g/cm³ (20 °C) Flash Pt: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003286 CAS Number: 4472-41-7 EINECS: 224-745-8 UN: 2265 ADR: 3, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...