Sodium dihydrogen phosphate monohydrate 90,0-102,0 Alkalimetric, EMSURE® ACS, Reag. Ph. Eur. zA, Supelco®

Supplier: MilliporeSigma

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

MQ300

1.06346.0500 1.06346.9012
CA1.06346.0500CS 1040 CAD
CA1.06346.0500 CA10754-004
Sodium dihydrogen phosphate monohydrate 90,0-102,0 Alkalimetric, EMSURE® ACS, Reag. Ph. Eur. zA, Supelco®
Sodium dihydrogen phosphate monohydrate

EMSURE® (formerly GR) is a premium grade of reagents for wet chemical analysis, adhering to ACS, ISO, and/or Reag. Ph. Eur. quality specifications.


1.06346.0500 replaces and is the same as SX0709-5. Part # change only, Same pack size, same specifications.

Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100
Density: 2.04 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
EINECS: 231-449-2
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Alkalimetric) 99.0 - 102.0%
Insoluble Matter ≤0.01%
pH-Value (5%; Water, 25 °C) 4.1 - 4.5
UV Absorption (280 nm; 1 mol/L; 1 cm; Water) ≤0.05
Chloride (Cl) ≤0.0005%
Sulfate (SO₄) ≤0.003%
Total Nitrogen (N) ≤0.001%
Heavy Metals (ACS) ≤0.0005%
Ca (Calcium) ≤0.005%
Fe (Iron) ≤0.0005%
K (Potassium) ≤0.005%
Corresponds to ACS, Reag. Ph. Eur.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR