Chloroform-[D₁] ≥99.8% (Isotopic) with TMS (0.03 vol.%) stabilized, MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Chloroform-D
, Trichloromethane-D1
, Chloroform-[²H₁]
Stabilization:
Stabilised with silver
M-Clarity™ quality level = MQ200
1.03296.0500
1.03296.0500
1.03296.0100
1.03296.0100
1.03296.0025
1.03296.0025
CA1.03296.0500EA
1119.94
CAD
CA1.03296.0500
CA1.03296.0025
EM1.03296.0025
EM1.03296.0100
CA1.03296.0100
EM1.03296.0500
Chloroform-[D₁] ≥99.8% (Isotopic) with TMS (0.03 vol.%) stabilized, MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Chloroform-[D1]
Formula:
CDCl₃ MW: 120.38 g/mol Boiling Pt: 60.8 °C (1013 hPa) Melting Pt: 60.8 °C (1013 hPa) Density: 1.5 g/cm³ (20 °C) |
MDL Number:
MFCD00000827 CAS Number: 865-49-6 UN: 1888 ADR: 6.1,III |
Specification Test Results
Deuteration degree | ≥ 99.8 % |
Water (NMR, H₂O) | ≤ 0.01 % |
Tetramethylsilane (NMR-spectrum) | 0.0003 |
Performance test (NMR-spectrum) | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...