Chloroform-[D₁] ≥99.8% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
1.02450.0100
1.02450.0500
CA1.02450.0100EA
223
CAD
CA1.02450.0100
CA1.02450.0025
CA1.02450.0500
Chloroform-[D₁] ≥99.8% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Chloroform-[D1]
MagniSolv™ deuterated solvents provide reliable results in the NMR-spectra with extremely low residual water, excellent chemical purity and the highest isotopic enrichment.
Formula:
CDCl₃ Boiling Pt: 60.8 °C (1013 hPa) Melting Pt: –64.1 °C Density: 1.5 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000827 CAS Number: 865-49-6 EINECS: 212-742-4 UN: 1888 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...