Ammonium acetate, EMSURE® zA, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Acetic acid ammonium salt

MQ300

1.01116.0500 1.01116.1000 1.01116.9050 1.01116.9025
CA1.01116.0500EA 845 CAD
CA1.01116.0500 CA1.01116.1000 CA1.01116.9050 EM1.01116.9025
Ammonium acetate, EMSURE® zA, Supelco®
Ammonium acetate

EMSURE® (formerly GR) is a premium grade of reagents for wet chemical analysis, adhering to ACS, ISO, and/or Reag. Ph.Eur quality specifications.


In North America 1.01116.0500 replaces but has a superior specification to AX1220-1
In North America 1.01116.1000 or 1.01116.5000 replaces but has a superior specification and different pack sizes to
AX1220-3
In North America 1.01116.9012 replaces but has a superior specification to AX1220-5
In North America 1.01116.9050 replaces but has a superior specification to AX1220-20

Formula: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: 114
Density: 1.17 g/cm3 at 20 °C
Flash Pt: 136
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR