MicroElute® Cycle-Pure & Gel Extraction Kit, Omega Bio-tek
Supplier: OMEGA BIO-TEK CA
D6294-00 D6294-02
CA101319-104EA 39.97 CAD
CA101319-104 CA101319-106 CA101319-300
MicroElute® Cycle-Pure & Gel Extraction Kit, Omega Bio-tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
MicroElute® Clean Up system, designed for rapid DNA clean up.
  • Elution volumes – 10-15 µL
  • Generation of highly concentrated DNA
  • 10 µg DNA binding capacity
  • Rapid isolation – less than 15 minutes
The E.Z.N.A.® family of products is an innovative system that radically simplifies the extraction and purification of nucleic acids from a variety of sources. The key to this system is the HiBind® matrix that specifically, but reversibly, binds DNA or RNA under optimized conditions allowing proteins and other contaminants to be removed. Nucleic acids are easily eluted with deionized water or a low salt buffer.
The MicroElute® Clean Up system, designed for rapid DNA clean up, includes:
• MicroElute® Cycle-Pure – for direct purification of double or single stranded PCR products (100 bp – 10 kb) from amplification reactions
• MicroElute® Gel Extraction – for extraction of DNA fragments from standard, or low-melt agarose gels in TAE (Tris-acetate/EDTA) or TBE (Tris-borate/EDTA) buffer