E.Z.N.A.® SP Plant DNA Kit , Omega Bio-tek®
Supplier: OMEGA BIO-TEK CA
D5511-00 D5511-02 D5511-01
CA101319-074EA 42.23 CAD
CA101319-074 CA101319-280 CA101319-368
E.Z.N.A.® SP Plant DNA Kit , Omega Bio-tek®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The E.Z.N.A.® SP Plant DNA Kit is specially designed for the rapid and reliable isolation of high-quality total cellular DNA from variety of plant species and tissues
Up to 100mg of fresh sample, or 30mg of dry sample can be processed in less than 40 minutes. The optimized procedure incorporates the homogenizer spin column, a unique filter filtration, and a homogenization column that can efficiently remove cell debris and improve the sample handling following the sample lysis. Purified DNA is suitable for most downstream applications including PCR, restriction enzyme digestion, and hybridization techniques.