E.Z.N.A.® FastFilter Plasmid Midi Kit, Omega Bio-tek®

Supplier: OMEGA BIO-TEK CA

D6905-03 D6905-04 D6905-00
CA101318-894EA 433.63 CAD
CA101318-894 CA101318-912 CA101414-400
E.Z.N.A.® FastFilter Plasmid Midi Kit, Omega Bio-tek®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

The E.Z.N.A.® FastFilter Plasmid Midi Kit isolates plasmid DNA from up to 50mL of bacterial culture in under 40 minutes.

Order Now

SPECIFICATIONS

Description FastFilter Plasmid Midi Kit
Environmentally Preferable
Application
Target molecule Plasmid
Refine target molecule Plasmid
Sample type Bacteria
Method / Format Column (silica-based)
Prep size Midi

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR